siRNA Details
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033321.3 | NP_201578.2 | 1 | 189 | ||
GenBank mRNA | AF333331.1 | - | 1 | 195 | ||
AK134662.1 | - | 1 | 149 | |||
BC138189.1 | - | 1 | 189 | 8861208 | ||
BC145181.1 | - | 1 | 189 | 9053732 | ||
JX570808.1 | - | 1 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
