siRNA Details
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053159.3 | NP_444389.2 | 5 | 643 | ||
GenBank mRNA | AB049633.1 | - | 5 | 524 | ||
AK002509.1 | - | 5 | 618 | |||
AK008592.1 | - | 5 | 673 | |||
AK054185.1 | - | 2 | 78 | |||
AK077857.1 | - | 5 | 643 | |||
AK145687.1 | - | 5 | 673 | |||
AK169272.1 | - | 5 | 644 | |||
BC011128.1 | - | 5 | 612 | 4187019 | ||
BC016586.1 | - | 5 | 516 | 2616397 | ||
BC034877.1 | - | 5 | 609 | 1547459 | ||
KC508086.1 | - | 5 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
