siRNA Details
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053159.3 | NP_444389.2 | 5 | 636 | ||
GenBank mRNA | AB049633.1 | - | 5 | 517 | ||
AK002509.1 | - | 5 | 611 | |||
AK008592.1 | - | 5 | 666 | |||
AK054185.1 | - | 2 | 71 | |||
AK077857.1 | - | 5 | 636 | |||
AK145687.1 | - | 5 | 666 | |||
AK169272.1 | - | 5 | 637 | |||
BC011128.1 | - | 5 | 605 | 4187019 | ||
BC016586.1 | - | 5 | 509 | 2616397 | ||
BC034877.1 | - | 5 | 602 | 1547459 | ||
KC508086.1 | - | 5 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
