siRNA Details
Gene Symbol: Mrpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053161.2 | NP_444391.1 | 4 | 322 | ||
GenBank mRNA | AB049648.1 | - | 4 | 317 | ||
AK143575.1 | - | 4 | 351 | |||
BC030074.1 | - | 4 | 322 | 1514322 | ||
BC096568.1 | - | 4 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
