siRNA Details
Gene Symbol: Mrpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053162.2 | NP_444392.1 | 2 | 512 | ||
GenBank mRNA | AB049653.1 | - | 2 | 464 | ||
AK018724.1 | - | 2 | 464 | |||
AK170156.1 | - | 2 | 455 | |||
BC052086.1 | - | 2 | 479 | 5684412 | ||
BC056979.1 | - | 2 | 463 | 6814476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
