siRNA Details
Gene Symbol: Mrpl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053163.1 | NP_444393.1 | 2 | 841 | ||
GenBank mRNA | AK131597.1 | - | 2 | 833 | ||
AK131601.1 | - | 2 | 831 | |||
AK160778.1 | - | 2 | 842 | |||
BC009166.1 | - | 2 | 841 | 2811744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
