siRNA Details
Gene Symbol: Mrpl43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053164.3 | NP_444394.1 | 2 | 229 | ||
GenBank mRNA | AB049657.1 | - | 2 | 190 | ||
AK015357.1 | - | 2 | 210 | |||
AK076204.1 | - | 2 | 188 | |||
BC027676.1 | - | 2 | 169 | |||
BC048733.1 | - | 2 | 220 | 6512868 | ||
BC065812.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
