siRNA Details
Gene Symbol: Clec2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053165.5 | NP_444395.2 | 3 | 435 | ||
GenBank mRNA | AF350410.1 | - | 3 | 414 | ||
AK017207.1 | - | 3 | 431 | |||
AK090364.1 | - | 3 | 435 | |||
AK136426.1 | - | 4 | 690 | |||
BC021766.1 | - | 3 | 411 | 4238060 | ||
HQ713437.1 | - | 3 | 342 | |||
HQ875061.1 | - | 3 | 378 |
