siRNA Details
Gene Symbol: Clec2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053165.5 | NP_444395.2 | 4 | 553 | ||
GenBank mRNA | AF350410.1 | - | 4 | 532 | ||
AK017207.1 | - | 4 | 549 | |||
AK090364.1 | - | 4 | 553 | |||
AK136426.1 | - | 6 | 965 | |||
BC021766.1 | - | 4 | 529 | 4238060 |
