siRNA Details
Gene Symbol: Trim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF220033.1 | - | 1 | 164 | ||
BC138188.1 | - | 1 | 140 | 8861207 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF220033.1 | - | 1 | 164 | ||
BC138188.1 | - | 1 | 140 | 8861207 |