siRNA Details
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286387.1 | NP_001273316.1 | 9, 10 | 2275 | ||
NM_001286388.1 | NP_001273317.1 | 9, 10 | 2278 | |||
NM_053167.3 | NP_444397.2 | 9, 10 | 2278 | |||
GenBank mRNA | AK049907.1 | - | 9, 10 | 1992 | ||
AK129109.1 | - | 9, 10 | 1983 | |||
AK163123.1 | - | 9, 10 | 2012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
