siRNA Details
Gene Symbol: Trim34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030684.3 | NP_109609.2 | 2 | 522 | ||
GenBank mRNA | AF220139.1 | - | 2 | 419 | ||
AF220140.1 | - | 422 | ||||
AK089187.1 | - | 537 | ||||
AK171840.1 | - | 2 | 482 | |||
AK172135.1 | - | 2 | 482 |
Gene Symbol: Trim34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243916.1 | NP_001230845.1 | 1 | 263 | ||
GenBank mRNA | BC145515.1 | - | 1 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
