siRNA Details
Gene Symbol: Med15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040683.2 | NP_001035773.1 | 2 | 245 | ||
NM_001285884.1 | NP_001272813.1 | 2 | 245 | |||
NM_001285886.1 | NP_001272815.1 | 4 | 399 | |||
NM_033609.3 | NP_291087.2 | 2 | 245 | |||
GenBank mRNA | AF328770.1 | - | 2 | 121 | ||
AK038915.1 | - | 2 | 208 | |||
AK052231.1 | - | 2 | 211 | |||
AK131186.1 | - | 2 | 186 | |||
AK142224.1 | - | 2 | 211 | |||
AK160772.1 | - | 2 | 211 | |||
AK166522.1 | - | 4 | 399 | |||
AK169904.1 | - | 2 | 246 | |||
AK171628.1 | - | 2 | 189 | |||
BC054779.1 | - | 2 | 195 | 6812541 | ||
BY201797.1 | - | 2 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
