siRNA Details
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053177.1 | NP_444407.1 | 6, 7 | 853 | ||
GenBank mRNA | AF302009.1 | - | 6, 7 | 850 | ||
AK028385.1 | - | 6, 7 | 830 | |||
AK034471.1 | - | 6, 7 | 864 | |||
AK038594.1 | - | 6, 7 | 866 | |||
AK049606.1 | - | 6, 7 | 854 | |||
BC005651.1 | - | 6, 7 | 853 | 3257440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
