siRNA Details
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052976.3 | NP_443208.1 | 17 | 2015 | ||
GenBank mRNA | AK028822.1 | - | 17 | 1499 | ||
AK031419.1 | - | 17 | 2008 | |||
AK036038.1 | - | 17 | 2016 | |||
AK082551.1 | - | 17 | 1655 | |||
AK087469.1 | - | 17 | 1479 | |||
AK178859.1 | - | 3 | 168 | |||
BC004845.1 | - | 18 | 1613 | 2650009 |
