siRNA Details
Gene Symbol: C1galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052993.3 | NP_443719.3 | 3 | 828 | ||
GenBank mRNA | AF157962.1 | - | 3 | 788 | ||
BC025899.1 | - | 2 | 451 | |||
BC064767.1 | - | 3 | 719 | 30077842 | ||
BG077752.2 | - | 1 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
