siRNA Details
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053183.2 | NP_444413.1 | 7 | 1106 | ||
GenBank mRNA | AF334104.1 | - | 7 | 1107 | ||
AK030239.1 | - | 8 | 1178 | |||
AK041281.1 | - | 7 | 1005 | |||
BC049906.1 | - | 7 | 1107 | |||
BC055440.1 | - | 7 | 1101 | |||
BC075714.1 | - | 7 | 1059 | |||
BC090996.1 | - | 7 | 1126 | 6757408 |
