siRNA Details
Gene Symbol: Hecw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081348.3 | NP_001074817.3 | 3 | 567 | ||
GenBank mRNA | AB083710.1 | - | 1 | 70 | ||
AK047678.1 | - | 3 | 502 | |||
AK051569.1 | - | 2 | 388 | |||
AK053694.1 | - | 2 | 512 | |||
AK084246.1 | - | 2 | 388 | |||
BB606770.2 | - | 2 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
