siRNA Details
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019791.2 | NP_062765.1 | 6 | 1726 | ||
GenBank mRNA | AB029448.1 | - | 6 | 1726 | ||
AF319975.1 | - | 5 | 1496 | |||
AK017275.1 | - | 6 | 1644 | |||
AK049660.1 | - | 6 | 1677 | |||
AK087937.1 | - | 6 | 1677 | |||
AK132248.1 | - | 6 | 1665 | |||
AK135452.1 | - | 6 | 1676 | |||
AK145872.1 | - | 6 | 1646 | |||
AK146867.1 | - | 6 | 1676 | |||
AK146908.1 | - | 6 | 1676 | |||
AK150876.1 | - | 3 | 665 | |||
AK195822.1 | - | 3 | 138 | |||
AK220150.1 | - | 7 | 1583 | |||
BC016438.1 | - | 6 | 1642 | 4480947 | ||
BC031461.1 | - | 6 | 1666 | 4018875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
