siRNA Details
Gene Symbol: Ugt1a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145079.3 | NP_659545.2 | 261 | |||
GenBank mRNA | AK162514.1 | - | 257 | |||
AY227197.1 | - | 1 | 229 | |||
U09930.1 | - | 1 | 143 | |||
U16818.1 | - | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
