siRNA Details
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168256.1 | NP_001161728.1 | 2 | 263 | ||
NM_001168257.1 | NP_001161729.1 | 2 | 263 | |||
NM_144805.2 | NP_659054.2 | 2 | 131 | |||
GenBank mRNA | BC019416.1 | - | 2 | 93 | 5151159 | |
BY104106.1 | - | 2 | 118 | |||
BY279100.1 | - | 2 | 263 | |||
CT010303.1 | - | 1 | 61 | |||
CV561368.1 | - | 2 | 205 | |||
CX564997.1 | - | 2 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
