siRNA Details
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026122.4 | NP_080398.1 | 4, 5 | 301 | ||
NM_175074.2 | NP_778249.1 | 4, 5 | 301 | |||
GenBank mRNA | AK002970.1 | - | 4, 5 | 220 | ||
AK003272.1 | - | 4, 5 | 249 | |||
AK153223.1 | - | 4, 5 | 261 | |||
BC005693.1 | - | 4, 5 | 237 | 3597594 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
