siRNA Details
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110146.1 | NP_001103616.1 | 10 | 903 | ||
NM_021400.3 | NP_067375.3 | 11 | 3297 | |||
GenBank mRNA | AB034730.1 | - | 12 | 2796 | ||
AK132597.1 | - | 5 | 1043 | |||
AK149469.1 | - | 10 | 904 | |||
BC130021.1 | - | 9 | 867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
