siRNA Details
Gene Symbol: Ptges2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133783.2 | NP_598544.2 | 4 | 935 | ||
GenBank mRNA | AK002798.1 | - | 4 | 559 | ||
AK050616.1 | - | 4 | 561 | |||
AK182234.1 | - | 4 | 294 | |||
BC004846.1 | - | 4 | 567 | 2650123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
