siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 12 | 1233 | ||
GenBank mRNA | AK004345.1 | - | 2 | 114 | ||
AK034867.1 | - | 11 | 1094 | |||
AK161445.1 | - | 10 | 1072 | |||
AK178753.1 | - | 2 | 108 | |||
AK183694.1 | - | 2 | 142 | |||
AK214695.1 | - | 4 | 286 | |||
BC012436.1 | - | 10 | 761 | |||
BC018294.1 | - | 12 | 1183 | 4021243 | ||
DV648379.1 | - | 4 | 292 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107880, XM_006237026, XM_006237025, XM_006237024.
Gene Ontology Categories:
