siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 4 | 600 | ||
GenBank mRNA | AK034867.1 | - | 4 | 547 | ||
AK178962.1 | - | 2 | 99 | |||
AK181484.1 | - | 2 | 91 | |||
AK191101.1 | - | 3 | 222 | |||
AK207311.1 | - | 1 | 47 | |||
AK210125.1 | - | 3 | 222 | |||
AK218803.1 | - | 3 | 222 | |||
BC012436.1 | - | 2 | 128 | |||
BC018294.1 | - | 4 | 550 | 4021243 | ||
CD805346.1 | - | 2 | 146 |
