siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 11 | 1106 | ||
GenBank mRNA | AK161445.1 | - | 9 | 945 | ||
AK183694.1 | - | 1 | 15 | |||
AK214695.1 | - | 3 | 159 | |||
BC012436.1 | - | 9 | 634 | |||
BC018294.1 | - | 11 | 1056 | 4021243 | ||
CD805346.1 | - | 9 | 652 | |||
DV648379.1 | - | 3 | 165 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107880, XM_006237026, XM_006237025, XM_006237024.
Gene Ontology Categories:
