siRNA Details
Gene Symbol: C78339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033192.2 | NP_001028364.1 | 3 | 958 | ||
GenBank mRNA | AK090371.1 | - | 3 | 342 | ||
AK141282.1 | - | 3 | 959 | |||
AK171039.1 | - | 4 | 859 | |||
AK218359.1 | - | 2 | 237 |

More Information
Other Species Targeted

Gene Ontology Categories:
