siRNA Details
Gene Symbol: Efhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028889.2 | NP_083165.1 | 4 | 689 | ||
GenBank mRNA | AK016491.1 | - | 4 | 689 | ||
AK045911.1 | - | 4 | 691 | |||
AK144175.1 | - | 4 | 692 | |||
AK171846.1 | - | 4 | 691 | |||
BC019531.1 | - | 4 | 675 | 4459208 | ||
BC085088.1 | - | 4 | 674 | 6467625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
