siRNA Details
Gene Symbol: AI597479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133818.1 | NP_598579.1 | 2 | 202 | ||
GenBank mRNA | AK081372.1 | - | 2 | 193 | ||
BC006931.1 | - | 2 | 202 | 3598975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
