siRNA Details
Gene Symbol: Nucks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145804.1 | NP_001139276.1 | 5 | 517 | ||
NM_175294.3 | NP_780503.2 | 5 | 517 | |||
GenBank mRNA | AB049825.1 | - | 5 | 496 | ||
AK078816.1 | - | 5 | 441 | |||
BC039951.1 | - | 5 | 429 | 4188558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
