siRNA Details
Gene Symbol: Cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030131.3 | NP_084407.1 | 4 | 353 | ||
GenBank mRNA | AK021301.1 | - | 4 | 307 | ||
AK144157.1 | - | 4 | 314 | |||
AK165179.1 | - | 4 | 299 | |||
AK166153.1 | - | 4 | 297 | |||
BC065395.1 | - | 4 | 297 | 876056 |

More Information
Other Species Targeted

Gene Ontology Categories:
