siRNA Details
Gene Symbol: Celrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045543.1 | - | 44 | |||
AK138346.1 | - | 1 | 112 | |||
BE654424.1 | - | 1 | 22 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
