siRNA Details
Gene Symbol: Pid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003948.2 | NP_001003948.2 | 2 | 361 | ||
GenBank mRNA | AK035542.1 | - | 2 | 236 | ||
AK083256.1 | - | 2 | 281 | |||
AK150971.1 | - | 2 | 299 | |||
AK151267.1 | - | 2 | 331 | |||
AK164328.1 | - | 2 | 328 | |||
BC080290.1 | - | 2 | 307 | 6816865 | ||
BY004446.1 | - | 2 | 361 |
