siRNA Details
Gene Symbol: Mfsd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133829.2 | NP_598590.2 | 2 | 1397 | ||
NM_178081.4 | NP_835182.1 | 2 | 1397 | |||
GenBank mRNA | AB247936.1 | - | 1 | 1012 | ||
AK035995.1 | - | 2 | 1398 | |||
AK046866.1 | - | 2 | 1541 | |||
AK171946.1 | - | 2 | 1438 | |||
BC017534.1 | - | 1 | 506 | 4488390 | ||
BC029324.1 | - | 1 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
