siRNA Details
Gene Symbol: Fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178887.4 | NP_849218.2 | 7 | 1376 | ||
GenBank mRNA | AK046338.1 | - | 7 | 1338 | ||
AK084379.1 | - | 8 | 1379 | |||
BC060634.1 | - | 7 | 1376 | 6848779 |
