siRNA Details
Gene Symbol: Cep152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081091.1 | NP_001074560.1 | 26 | 4284 | ||
GenBank mRNA | AK028650.1 | - | 1 | 474 | ||
AK173060.1 | - | 26 | 4284 | |||
BC026366.1 | - | 6 | 1012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
