siRNA Details
Gene Symbol: Hrh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133849.3 | NP_598610.1 | 2 | 528 | ||
GenBank mRNA | AK140374.1 | - | 2 | 533 | ||
AY044153.2 | - | 2 | 355 | |||
AY142145.1 | - | 2 | 499 | |||
BC132201.1 | - | 2 | 295 | 40130478 | ||
BC145125.1 | - | 2 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
