siRNA Details
Gene Symbol: Wdr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181400.3 | NP_852065.2 | 6, 7 | 1594 | ||
GenBank mRNA | BC040337.1 | - | 6, 7 | 1455 | 3586908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
