siRNA Details
Gene Symbol: Arfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081093.2 | NP_001074562.1 | 7, 8 | 1020 | ||
NM_001293801.1 | NP_001280730.1 | 6, 7 | 924 | |||
GenBank mRNA | AK160182.1 | - | 2, 3 | 349 | ||
BC132291.1 | - | 6, 7 | 833 | 40130568 | ||
BC144758.1 | - | 6, 7 | 833 | 9053306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
