siRNA Details
Gene Symbol: Cyp2j11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004141.2 | NP_001004141.2 | 2 | 354 | ||
GenBank mRNA | AK143983.1 | - | 2 | 336 | ||
AW260720.1 | - | 2 | 354 | |||
BC080831.1 | - | 2 | 330 | 30469795 |
