siRNA Details
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081557.3 | NP_001075026.1 | 3 | 236 | ||
NM_001195565.1 | NP_001182494.1 | 3 | 236 | |||
GenBank mRNA | AK009988.1 | - | 2 | 136 | ||
AK034038.1 | - | 3 | 173 | |||
AK076589.1 | - | 2 | 49 | |||
AK144150.1 | - | 3 | 206 | |||
AK158710.1 | - | 3 | 237 | |||
BC150740.1 | - | 2 | 85 | 9087651 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001195559, NM_001195560, BF290447, BC106025.
Gene Ontology Categories:
