siRNA Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285872.1 | NP_001272801.1 | 10 | 1134 | ||
NM_001285874.1 | NP_001272803.1 | 9 | 1076 | |||
NM_001285875.1 | NP_001272804.1 | 9 | 1026 | |||
NM_001285877.1 | NP_001272806.1 | 7 | 770 | |||
NM_133880.3 | NP_598641.2 | 10 | 1123 | |||
GenBank mRNA | AK075918.1 | - | 2 | 66 | ||
AK144090.1 | - | 10 | 1086 | |||
AK145092.1 | - | 9 | 977 | |||
AK156774.1 | - | 9 | 1077 | |||
AK205204.1 | - | 2 | 137 | |||
BC021890.1 | - | 9 | 1028 | 3598000 | ||
BC025495.1 | - | 10 | 1145 | 5310200 | ||
BC025938.1 | - | 9 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
