siRNA Details
Gene Symbol: Oas1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133893.3 | NP_598654.1 | 2 | 377 | ||
GenBank mRNA | AB067532.1 | - | 2 | 331 | ||
AK054465.1 | - | 2 | 345 | |||
AK136001.1 | - | 2 | 345 | |||
AY055829.1 | - | 2 | 339 | |||
BC052835.1 | - | 2 | 341 | 30048758 | ||
DV659301.1 | - | 2 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
