siRNA Details
Gene Symbol: AA792892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178894.4 | NP_849225.2 | 3 | 674 | ||
GenBank mRNA | AK053948.1 | - | 629 | |||
AK087752.1 | - | 698 | ||||
BC120516.1 | - | 3 | 674 | 40129439 | ||
BC125434.1 | - | 3 | 674 | 40129949 |
