siRNA Details
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133900.4 | NP_598661.1 | 7 | 1107 | ||
GenBank mRNA | AA003039.1 | - | 1 | 52 | ||
AK088865.1 | - | 7 | 1059 | |||
AK168048.1 | - | 5 | 715 | |||
AK217223.1 | - | 2 | 128 | |||
BC002251.1 | - | 5 | 678 | 3493291 | ||
BQ177662.1 | - | 5 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
