siRNA Details
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190733.1 | NP_001177662.1 | 9 | 812 | ||
NM_001190734.1 | NP_001177663.1 | 11 | 852 | |||
NM_178896.5 | NP_849227.1 | 10 | 917 | |||
GenBank mRNA | AK033559.1 | - | 10 | 862 | ||
AK076991.1 | - | 11 | 853 | |||
BC031793.1 | - | 1 | 41 | |||
BC060213.1 | - | 10 | 859 | |||
BC108346.1 | - | 10 | 819 | 6310392 | ||
BC137627.1 | - | 10 | 861 | 8860646 | ||
BC137628.1 | - | 10 | 861 | 8860647 | ||
BC144794.1 | - | 9 | 756 | 9053342 | ||
CO433728.1 | - | 5 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
