siRNA Details
Gene Symbol: Rita1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029096.3 | NP_083372.1 | 2 | 537 | ||
NM_133908.1 | NP_598669.1 | 3 | 632 | |||
GenBank mRNA | AK003581.1 | - | 2 | 518 | ||
BC021365.1 | - | 2 | 436 | 5030737 | ||
CX239969.1 | - | 3 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
