siRNA Details
Gene Symbol: Rab28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027295.2 | NP_081571.1 | 5 | 608 | ||
GenBank mRNA | AB232624.1 | - | 5 | 432 | ||
AK012286.1 | - | 5 | 611 | |||
AK159493.1 | - | 5 | 609 | |||
AK161391.1 | - | 5 | 489 | |||
AK215070.1 | - | 4 | 344 | |||
BC004580.1 | - | 5 | 608 | 3587008 |
