siRNA Details
Gene Symbol: Zfp282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146175.3 | NP_666287.3 | 6 | 1168 | ||
GenBank mRNA | AK030516.1 | - | 6 | 1076 | ||
AK088531.1 | - | 6 | 1167 | |||
AK148038.1 | - | 6 | 1154 | |||
BC025581.1 | - | 6 | 1070 | 5325029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
